Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Nucleocapsid protein, His Tag
Marke:  ACROBiosystems NUN-C5227-100UG
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16575174
  • 479.87 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
QHO62115.1
100 μg
Met 1 - Ala 419
Nucleocapsid protein,NP,Protein N,COVID-19
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
47.3 kDa
SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C5227) is expressed and secreted by human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1).
-20°C
Nucleocapsid protein
SARS-CoV-2
PBS, pH7.4
HEK293
Viral Proteins