Learn More
FMC63 scFv Mouse, FITC, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems FM3-FY45-200tests
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.Spezifikation
FMC63 scFv | |
Y45 | |
Hybridoma | |
RUO | |
-20°C, aviod light |
Monoclonal | |
FITC | |
200 Tests | |
Primary | |
IgG1 |