Learn More
ACROBiosystems Biotinylated Human FGL1 (64-312) Protein, Avitag™,Fc Tag (high sensitivity)
Biotinylated Protein
Marke:  ACROBiosystems FG1-H82F6-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17106321
  • 1847.47 € / 200 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Fibrinogen-like protein 1(FGL1) is also known as HP-041, Hepassocin, HFREP-1, LFIRE-1. The protective effect of fibrinogen-like protein 1 (FGL1) in liver injury has previously been reported. However, studies have shown that FGL1 may be a predictor of GC patients and a target for GC therapy. Immunocytochemical studies revealed that fgl1 selectively binds to defective spermatozoa in the cauda epididymidis. Northern blot analysis and in situ hybridization demonstrated the high expression of fgl1 in the principal cells of the proximal cauda epididymidis. Immunofluorescence analysis using mouse fibrotic lung tissues suggested that fibrotic regions showed increased expressions of Gtse1 and Fgl1, Gtse1 and Fgl1 are suggested to be novel targets for radiation-induced lung fibrosis.
This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 116.3 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
NP_004458.3
200 μg
Asp 64 - Ile 312
FGL1,Hepassocin,HP-041,HFREP-1,LFIRE-1,HFREP1
Precision Avi
Avi Tag and Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
116.3 kDa
Biotinylated Human FGL1 (64-312), Avitag,Fc Tag (FG1-H82F6) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08830-1).
-20°C
FGL1
Human
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
MABSol™ Biotin Labeled Proteins