Learn More
ACROBiosystems Cynomolgus IL-1 Rrp2 / IL-1 R6 Protein, His Tag
Marke: ACROBiosystems IL2-C52H5-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The Interleukin 1 receptor family (IL-1 R) comprises at least eleven members including IL-1 RI (IL-1 R1), IL-1 RII (IL-1 R2), IL-1 RAcP (IL‐1 R3), ST2 (T1/IL-1 R4), IL-18 Ra (IL-1 Rrp/IL-1 R5), IL-1 Rrp2 (IL-1 RL2/IL-1 R6), IL-18 Rb (AcPL/IL-1 R7), IL-1RAPL‐1 (TIGIRR‐2/IL‐1 R8), and TIGIRR-1 (IL-1 R9). All family members possess three immunoglobulin (Ig)-like domains in their extracellular region. Most members also have an intracellular TIR (Toll-like receptor/IL-1 receptor signaling) domain that is also conserved in the Toll-like receptor family. Related proteins, SIGIRR (single Ig domain-containing IL-1 R-related molecule) and IL-18BP, differ from the other members by having only one Ig domain.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Spezifikation
EHH61702.1 | |
1 mg | |
Asp 20 - Arg 335 | |
IL-1 R6,IL-1 Rrp2,IL1RL2,IL-1 RL2,IL1Rrp2,IL-1 Rrp2,IL1RRP2,IL-36 R | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
37.9 kDa | |
Cynomolgus IL-1 Rrp2, His Tag (IL2-C52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 335 (Accession # EHH61702.1). | |
-20°C | |
IL-1 Rrp2 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |