Learn More
ACROBiosystems Human IL-17 RC Protein, Fc Tag
MALS verified
Marke:  ACROBiosystems ILC-H5259-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17106441
  • 2639.24 € / 1 Milligramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Interleukin-17 receptor C (IL-17 RC), also known as IL17Rhom, IL-17RL, is the receptor for IL17A and IL17F homodimers as part of a heterodimeric complex with IL17RA. IL-17A activity is inhibited by IL-17RA, IL-17F is inhibited by IL-17RC, and a combination of soluble IL-17RA/IL-17RC receptors is required for inhibition of the IL-17F/IL-17A activity. Furthermore, activation of IL17RC can lead to the induction of expression of inflammatory chemokines and cytokines such as CXCL1.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 75.9 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_703190.2
1 mg
Leu 21 - Ala 465
IL-17 RC,IL-17 RC,IL17Rhom,IL-17 receptor C,IL-17 RL,ZcytoR14
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
75.9 kDa
Human IL-17 RC, Fc Tag (ILC-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Ala 465 (Accession # NP_703190.2).
-20°C
IL-17 RC
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens