missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human IL-5 Protein, His, Avitag™

Biotinylated Protein

Marke:  ACROBiosystems IL5-H82Q5-200ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 17116421

  • 1847.47 € / 200 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Interleukin 5 (IL5) is an interleukin produced by type-2 T helper cells and mast cells. IL-5 is a 115-amino acid (in human, 133 in the mouse) -long TH2 cytokine that is part of the hematopoietic family. Unlike other members of this cytokine family (namely interleukin 3 and GM-CSF), this glycoprotein in its active form is a homodimer. Interleukin-5 has long been associated with the cause of several allergic diseases including allergic rhinitis and asthma, wherein a large increase in the number of circulating, airway tissue, and induced sputum eosinophils have been observed. Given the high concordance of eosinophils and, in particular, allergic asthma pathology, it has been widely speculated that eosinophils have an important role in the pathology of this disease. Drugs that target IL-5 are mepolizumab and reslizumab.

This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 16.7 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

16.7 kDa
Biotinylated Human IL-5, His,Avitag (IL5-H82Q5) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession # P05113-1).
-20°C
IL-5
Human
PBS,pH7.3
HEK293
MABSol™ Biotin Labeled Proteins
200 μg
Ile 20 - Ser 134
IL-5,TRF,IL5,Interleukin-5
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote