Learn More
ACROBiosystems Human LIGHT / TNFSF14 Protein, His Tag (active trimer)
MALS verified
Marke:  ACROBiosystems LIT-H5242-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17116461
  • 443.87 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Tumor necrosis factor ligand superfamily member 14(LIGHT) is a tumor necrosis factor (TNF) superfamily ligand that regulates T cell immune responses by signaling through the herpes virus entry mediator (HVEM) and the lymphotoxin beta receptor (LTbetaR). LIGHT has emerged as a potent initiator of T cell co-stimulation signals effecting CTL-mediated tumor rejection, allograft rejection and graft versus host disease. Constitutive expression of LIGHT leads to tissue destruction and autoimmune-like disease syndromes.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 58.3 kDa. The protein migrates as 55-65 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP 003798.2
100 μg
Asp 74 - Val 240
CD258 antigen,CD258,HVEM-L,LIGHT,LTg,TNFSF14,TR2,HVEML
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
58.3 kDa
Human LIGHT, His Tag (active trimer) (MALS verified) (LIT-H5242) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1) trimer Design.
-20°C
LIGHT
Human
PBS, pH7.4
HEK293
Immune Checkpoint Proteins