Learn More
ACROBiosystems Anti TIGIT ELISA Kit
For a rapid quantification of Anti-TIGIT antibodies through antigen binding
Marke:  ACROBiosystems TAT-K004-96tests
Weitere Details : Netto-Gewicht : 0.10000kg
Artikelnummer. 17116811
  • 685.98 € / 96 Tests
Voraussichtliches Versanddatum: 01-10-2021
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
TIGIT has been shown to be over expressed on tumor antigen-specific CD8+ T cells and CD8+ tumor infiltrating lymphocytes (TIL). Blockade of TIGIT led to increased cell proliferation, cytokine production, and degranulation of TA-specific CD8+ T cells and TIL CD8+ T cells, which now make it be considered as an immune checkpoint. Recently, studies have shown anti-TIGIT antibody is promising to become a cancer immunotherapy. Our ELISA kit is designed to help the discovery of anti-TIGIT therapeutic antibody.
Spezifikation
Spezifikation
ELISA Assay Kit
2°C to 8°C, aviod light
RT
This assay kit employs a standard Indirect-ELISA format, which provides a rapid quantification of Anti-TIGIT antibodies through antigen binding. The Kit includes of Streptavidin pre-coated plate, the Biotinylated TIGIT protein, a human anti-TIGIT antibody
96 Tests