Learn More
ACROBiosystems Mouse CD3 epsilon&CD3 delta Heterodimer Protein, His Tag&Flag Tag
MALS verified
Marke:  ACROBiosystems CDD-M5219-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17126221
  • 455.87 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Mouse CD3E&CD3D Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of CD3E and CD3D, has a calculated MW of 15.1 kDa (CD3E) and 14.3 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3D is fused wit
Spezifikation
Spezifikation
NP_031674.1 (CD3E) and NP_038515.3 (CD3D)
100 μg
Asp 23 - Asp 108 (CD3E) and Phe 22 - Ala 105 (CD3D)
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon
Unconjugated
His Tag and Flag Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
15.1 kDa (CD3E) and 14.3 kDa (CD3D)
Mouse CD3E&CD3D; Heterodimer Protein, His Tag&Flag; Tag (MALS verified) (CDD-M5219) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 108 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P22646-1 (CD3E) and P04235-1 (CD3D)).
-20°C
CD3E and CD3D
Mouse
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens