Learn More
ACROBiosystems Mouse NOTCH1 Protein, His Tag
Marke: ACROBiosystems NO1-M52H8-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
NOTCH1 Interacts with DNER, DTX1, DTX2 and RBPJ/RBPSUH. Also interacts with MAML1, MAML2 and MAML3 which act as transcriptional coactivators for NOTCH1. The NOTCH1 intracellular domain interacts with SNW1; the interaction involves multimerized NOTCH1 NICD and is implicated in a formation of an intermediate preactivation complex which associates with DNA-bound CBF-1/RBPJ. The activated membrane-bound form interacts with AAK1 which promotes NOTCH1 stabilization. Functions as a receptor for membrane-bound ligands Jagged-1 (JAG1), Jagged-2 (JAG2) and Delta-1 (DLL1) to regulate cell-fate determination. Involved in the maturation of both CD4+ and CD8+ cells in the thymus. Important for follicular differentiation and possibly cell fate selection within the follicle. During cerebellar development, functions as a receptor for neuronal DNER and is involved in the differentiation of Bergmann glia.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.4 kDa. The protein migrates as 58-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_032740.3 | |
100 μg | |
Ala 19 - Gln 526 | |
NOTCH1,Notch 1,hN1,TAN1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
55.4 kDa | |
Mouse NOTCH1, His Tag (NO1-M52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 526 (Accession # Q01705-1). | |
-20°C | |
NOTCH1 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |