Learn More
ACROBiosystems Anti Her2 ELISA Kit
For rapid quantification of Anti-Her2 antibodies through antigen binding
Marke:  ACROBiosystems TAH-K005-96tests
Weitere Details : Netto-Gewicht : 0.10000kg
Artikelnummer. 17126791
  • 685.98 € / 96 Tests
Voraussichtliches Versanddatum: 01-10-2021
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description
Description
Her2 is a member of the human epidermal growth factor receptor family. In recent years, it has become an important biomarker and target of therapy for oncology patients. Although Her2 has been identified as a target of therapy for years and there are a few anti-Her2 therapeutic antibodies on the market, the heat of research regarding this target remains on. ACRO anti Her2 ELISA kit is intended for PK study as well as anti Her2 therapeutic screening.
Spécification
Spécification
ELISA Assay Kit
2°C to 8°C, aviod light
RT
This assay kit employs a standard Indirect-ELISA format, which provides a rapid quantification of Anti-Her2 antibodies through antigen binding. The Kit includes of Streptavidin pre-coated plate, the Biotinylated Her2 protein, a human anti-Her2 antibody st
96 Tests
Vidéos
FDS
Documents
Certificats