Learn More
ACROBiosystems ActiveMax™ Human CCL19 / MIP-3 beta Protein, Tag Free
Marke: ACROBiosystems CC9-H5215-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
C-C motif chemokine 19(CCL19) is as known as CK beta-11, ELC, MIP3B and SCYA19. May play a role not only in inflammatory and immunological responses but also in normal lymphocyte recirculation and homing. May play an important role in trafficking of T-cells in thymus, and T-cell and B-cell migration to secondary lymphoid organs. Binds to chemokine receptor CCR7. Recombinant CCL19 shows potent chemotactic activity for T-cells and B-cells but not for granulocytes and monocytes. Binds to atypical chemokine receptor ACKR4 and mediates the recruitment of beta-arrestin (ARRB1/2) to ACKR4.
This protein carries no "e;tag"e;. The protein has a calculated MW of 8.8 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
8.8 kDa | |
ActiveMax™ Human CCL19 / MIP-3 beta, Tag Free (CC9-H5215) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Ser 98 (Accession # Q99731-1). | |
-20°C | |
CCL19 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |
1 mg | |
Gly 22 - Ser 98 | |
CCL19,MIP-3 beta,MIP-3-beta,ELC,MIP3B,SCYA19,EBI1 ligand chemokine | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |