Learn More
ACROBiosystems Human Siglec-1 / CD169 Protein, His Tag
Marke: ACROBiosystems SG1-H52H3-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Alternate names for sialoadhesin include siglec-1 and CD169 (cluster of differentiation 169). Since sialoadhesin binds sialic acids with its N-terminal IgV-domain, it is also a member of the SIGLEC family. It is defined as an I-type lectin, since it contains 17 immunoglobulin (Ig) domains (one variable domain and 16 constant domains), and thus also belongs to the immunoglobulin superfamily (IgSF). Since sialoadhesin binds sialic acids with its N-terminal IgV-domain, it is also a member of the SIGLEC family. Acts as an endocytic receptor mediating clathrin dependent endocytosis. Macrophage-restricted adhesion molecule that mediates sialic-acid dependent binding to lymphocytes, including granulocytes, monocytes, natural killer cells, B-cells and CD8 T-cells. Preferentially binds to alpha-2,3-linked sialic acid. Binds to SPN/CD43 on T-cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 175.1 kDa. The protein migrates as 200-220 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
175.1 kDa | |
Human Siglec-1, His Tag (SG1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Gln 1641 (Accession # Q9BZZ2-1). | |
-20°C | |
Siglec-1 | |
Human | |
MES,NaCl,pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
100 μg | |
Ser 20 - Gln 1641 | |
Siglec-1,CD169,SIGLEC1,SN | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |