Learn More
BCMA Neutralizing Mouse, Unconjugated, Monoclonal Antibody, recommended for neutralizing assay, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems BCA-M43-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
BCMA Neutralizing Monoclonal antibody specifically detects BCMA Neutralizing, recommended for neutralizing assay.Spezifikation
BCMA Neutralizing | |
Unconjugated | |
100 μg | |
Primary | |
IgG1 |
Monoclonal | |
Human (HEK293) | |
RUO | |
-20°C |