missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleocapsid protein Mouse anti-SARS-CoV-2, Unconjugated, Clone: AS47, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems NUN-S47A1-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Nucleocapsid protein Monoclonal antibody specifically detects Nucleocapsid protein in SARS-CoV-2 samples.Spezifikation
Nucleocapsid protein | |
AS47 | |
Human (HEK293) | |
RUO | |
SARS-CoV-2 | |
IgG1 |
Monoclonal | |
Unconjugated | |
100 μg | |
Primary | |
-20°C |