missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human RNF43 Protein, Fc Tag

MALS verified

Marke:  ACROBiosystems RN3-H5256-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 17146651

  • 419.88 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

E3 ubiquitin-protein ligase that acts as a negative regulator of the Wnt signaling pathway by mediating the ubiquitination, endocytosis and subsequent degradation of Wnt receptor complex components Frizzled. Acts on both canonical and non-canonical Wnt signaling pathway. Along with RSPO2 and ZNRF3, constitutes a master switch that governs limb specification (By similarity)

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

43.5 kDa
Human RNF43, Fc Tag (RN3-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 44 - Tyr 197 (Accession # Q68DV7-1).
-20°C
RNF43
Human
PBS, pH7.4
HEK293
Enzymes
100 μg
Gln 44 - Tyr 197
RING finger protein 43,RING-type E3 ubiquitin transferase RNF43
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote