Learn More
Spike S1 protein Mouse anti-SARS-CoV-2, Unconjugated, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems S1N-S59A1-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Spike S1 protein Monoclonal antibody specifically detects Spike S1 protein in SARS-CoV-2 samples.Spezifikation
Spike S1 protein | |
Unconjugated | |
100 μg | |
Primary | |
-20°C |
Monoclonal | |
Hybridoma | |
RUO | |
SARS-CoV-2 | |
IgG1 |