Learn More
FMC63 scFv Mouse, Unconjugated, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems FM3-Y45P1-1000tests
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.Spezifikation
FMC63 scFv | |
Y45 | |
Human (HEK293) | |
RUO | |
-20°C |
Monoclonal | |
Unconjugated | |
1000 Tests | |
Primary | |
IgG1 |