Learn More
ACROBiosystems Human IL-8 / CXCL8 Protein, Fc Tag
Marke:  ACROBiosystems IL8-H5258-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17156421
  • 419.88 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Interleukin 8 (IL8 or chemokine (C-X-C motif) ligand 8, CXCL8) is a chemokine produced by macrophages and other cell types such as epithelial cells, airway smooth muscle cells and endothelial cells. There are many receptors on the surface membrane capable of binding IL-8, the most frequently studied types are the G protein-coupled serpentine receptors CXCR1 and CXCR2. Expression and affinity for IL-8 differs between the two receptors (CXCR1 > CXCR2). IL-8(6-77) has a 5-10-fold higher activity on neutrophil activation, IL-8(5-77) has increased activity on neutrophil activation and IL-8(7-77) has a higher affinity to receptors CXCR1 and CXCR2 as compared to IL-8(1-77), respectively.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 34.8 kDa. The protein migrates as 37-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_000575.1
100 μg
Ser 28 - Ser 99
CXCL8,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
34.8 kDa
Human IL-8, Fc Tag (IL8-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).
-20°C
IL-8
Human
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5
HEK293
Cytokines and Growth Factors