Learn More
ACROBiosystems Biotinylated Human SIRP alphaV8 / CD172a Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems SI8-H82W6-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Tyrosine-protein phosphatase non-receptor type substrate 1 (SHPS1) is also known as CD172 antigen-like family member A (CD172a), Macrophage fusion receptor, MyD-1 antigen, Signal-regulatory protein alpha (SIRPA or SIRP alpha) or p84, is a member of the SIRP family, and also belongs to the immunoglobulin superfamily. SIRP alpha is Ubiquitous and highly expressed in brain. SIRPA / CD172a is immunoglobulin-like cell surface receptor for CD47 and acts as docking protein and induces translocation of PTPN6, PTPN11 and other binding partners from the cytosol to the plasma membrane. SIRPA / SHPS-1 supports adhesion of cerebellar neurons, neurite outgrowth and glial cell attachment and may play a key role in intracellular signaling during synaptogenesis and in synaptic function By similarity. SIRPA / MyD1 involved in the negative regulation of receptor tyrosine kinase-coupled cellular responses induced by cell adhesion, growth factors or insulin and mediates negative regulation of phagocytosis, mast cell activation and dendritic cell activation. CD47 binding prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
ATD50864.1 | |
200 μg | |
Glu 31 - Arg 369 (S44L, S50T, I52T, H54R, V57A) | |
SHPS1,SIRPA,CD172A,BIT,MFR,MYD1,P84,PTPNS1 | |
Unconjugated | |
His Tag and Avi Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>90% as determined by reduced SDS-PAGE |
40.8 kDa | |
Biotinylated Human SIRP alpha V8, His,Avitag (SI8-H82W6) is expressed from human 293 cells (HEK293). It contains AA Glu 31- Arg 369 (S44L, S50T, I52T, H54R, V57A) (Accession # ATD50864.1). | |
-20°C | |
SIRP alphaV8 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |