missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S protein RBD Potent Neutralizing Mouse anti-SARS-CoV-2, Unconjugated, Clone: AM128, Chimeric mAb, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems SPD-M128-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
S protein RBD Potent Neutralizing Monoclonal antibody specifically detects S protein RBD Potent Neutralizing in SARS-CoV-2 samples.Spezifikation
S protein RBD Potent Neutralizing | |
AM128 | |
Human (HEK293) | |
RUO | |
SARS-CoV-2 | |
IgG1 |
Monoclonal | |
Unconjugated | |
100 μg | |
Primary | |
-20°C |