Learn More
ACROBiosystems Human CD6 / TP120 Protein, His Tag
Marke:  ACROBiosystems CD6-H52H7-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17166131
  • 359.89 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
CD6 (Cluster of Differentiation 6) is a cell surface glycoprotein that is involved in T cell activation and a member of the scavenger receptor cysteine rich protein superfamily (SRCRSF). CD6 is expressed at low levels on immature thymocytes and at high levels on mature thymocytes. The majority of peripheral blood T cells, a subset of B cells, and a subset of neuronal cells express CD6. CD6 appears to play a role as both a costimulatory molecule in T cell activation and as an adhesion receptor. Studies demonstrating a mitogenic effect for T cells with some CD6 specific monoclonal antibodies, in conjunction with either accessory cells or PMA and anti-CD2 mAb, support the concept of CD6 as a co stimulatory molecule.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.5 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_006716.3
100 μg
His 18 - Glu 398
CD6,TP120,T12
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
42.5 kDa
Human CD6, His Tag (CD6-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 18 - Glu 398 (Accession # P30203-1).
-20°C
CD6
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens