Learn More
ACROBiosystems Biotinylated Mouse CD3 epsilon&CD3 delta Heterodimer Protein, His, Avitag™&Tag Free
Biotinylated Protein
Marke: ACROBiosystems CDD-M82W5-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Biotinylated Mouse CD3E&CD3D Heterodimer Protein, His,Avitag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 17.3 kDa (CD3E) and 13.3 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag and followed by an Avi tag (AviSpezifikation
17.3 kDa (CD3E) and 13.3 kDa (CD3D) | |
MABSol™ Biotinylated Mouse CD3E&CD3D; Heterodimer Protein, His,AviTag&Tag; Free (CDD-M82W5) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 108 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P22646-1 (CD3E) and P04235-1 (CD3D)). It is the biotinylated form of Mouse CD3 epsilon and CD3 delta Protein. | |
-20°C | |
CD3E and CD3D | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
200 μg | |
Asp 23 - Asp 108 (CD3E) and Phe 22 - Ala 105 (CD3D) | |
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |