Learn More
ACROBiosystems Human NKG2D / CD314 Protein, Mouse IgG2a Fc Tag
Marke:  ACROBiosystems NKD-H5259-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17166541
  • 359.89 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
NKG2D is a transmembrane protein belonging to the CD94/NKG2 family of C-type lectin-like receptors, also known as KLRK1, CD314, D12S2489E, KLR and killer cell lectin like receptor K1. NKG2D itself forms a homodimer whose ectodomains serve for ligand binding. NKG2D is a major recognition receptor for the detection and elimination of transformed and infected cells as its ligands are induced during cellular stress, either as a result of infection or genomic stress such as in cancer. In NK cells, NKG2D serves as an activating receptor, which itself is able to trigger cytotoxicity. The function of NKG2D on CD8+ T cells is to send co-stimulatory signals to activate them.
This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_031386.2 (T72A)
100 μg
Ile 73 - Val 216
NKG2D,CD314,KLRK1,NK cell receptor D
Unconjugated
Mouse IgG Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
43.5 kDa
Human NKG2D, Mouse IgG2a Fc Tag (NKD-H5259) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).
-20°C
NKG2D
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Other Recombinant Proteins