Learn More
ACROBiosystems Biotinylated Human PVRIG Protein, Fc,Avitag™
HPLC-verified
Marke: ACROBiosystems PVG-H82F9-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Human PVRIG (poliovirus receptor related immunoglobulin domain-containing protein), also known as CD112 receptor (CD112R), is an approximately 34 kDa single transmembrane protein in the poliovirus receptor-like protein (PVR) family. The CD112R gene encodes a putative single transmembrane protein, which is composed of a single extracellular IgV domain, one transmembrane domain, and a long intracellular domain. Notably, the intracellular domain of phatases. The extracellular domain sequence of human and mouse CD112R have 65.3% similarity. CD112R may act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.9 kDa. The protein migrates as 50-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_076975.2 | |
200 μg | |
Thr 41 - Asp 171 | |
C7orf15,C7orf15MGC138295,CD112R,MGC104322,MGC138297,MGC2463,PVRIG,CD112 receptor | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
41.9 kDa | |
Biotinylated Human PVRIG, Fc,Avitag™ (HPLC verified) (PVG-H82F9) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1). | |
-20°C | |
PVRIG | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |