Learn More
ACROBiosystems Human Activin RIB / ACVR1B Protein, His Tag
Marke: ACROBiosystems ACB-H52H4-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Activin receptor type-1B (ACVR1B) is also known as Activin receptor-like kinase 4 (ALK-4), Serine/threonine-protein kinase receptor R2 (SKR2), which belongs to the protein kinase superfamily, TKL Ser/Thr protein kinase family and TGFB receptor subfamily. ACVR1B contains one GS domain and one protein kinase domain. The catalytic activity of ACVR1B is “ATP + [receptor-protein] = ADP + [receptor-protein] phosphate”. ACVR1B acts as a transducer of activin or activin like ligands (e.g., inhibin) signals. Activin binds to either ACVR2A or ACVR2B and then forms a complex with ACVR1B. These go on to recruit the R-SMADs SMAD2 or SMAD3. ACVR1B also transduces signals of nodal, GDF-1, and Vg1.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.3 kDa. The protein migrates as 16-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH00254 | |
100 μg | |
Ser 24 - Glu 126 | |
ACVR1b,ALK4,ACVRLK4,ACTR-IB,SKR2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
13.3 kDa | |
Human Activin RIB, His Tag (ACB-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Glu 126 (Accession # P36896-1). | |
-20°C | |
Activin RIB | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |