Learn More
ACROBiosystems Human IFN-alpha 1 Protein, His Tag
Marke: ACROBiosystems IFA-H52H9-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interferon alpha-1 is also known as IFN-alpha-1/13, Interferon alpha-D, LeIF D and IFNA1, belongs to the alpha / beta interferon family. Interferons alpha-1 and alpha-13 have identical protein sequences. Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase. IFN-alpha can either suppress or promote the development of autoimmune diabetes. It is likely that IFN-alpha plays a complex role in the etiology of type 1 diabetes.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.3 kDa. The protein migrates as 22-24 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
21.3 kDa | |
Human IFN-alpha 1, His Tag (IFA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 189 (Accession # P01562-1). | |
-20°C | |
IFN-alpha 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |
100 μg | |
Cys 24 - Glu 189 | |
IFN-alpha 1,IFNA1,IFNalpha 1,IFN-alpha-1,LeIF D | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |