Learn More
ACROBiosystems Human Kremen-1 protein, Fc Tag
Marke: ACROBiosystems KR1-H5253-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Kremens are high-affinity receptors for Dickkopf 1 (Dkk1) and regulate the Wnt/β-catenin signaling pathway by down-regulating the low-density lipoprotein receptor-related protein 6 (LRP6). Kremen1 is considered as one component of a membrane complex modulating canonical Wnt signalling through LRP6 in vertebrates. Additionally, KREMEN1 (KRM1) has also been identified as a functional receptor for Coxsackievirus A10 (CV-A10), a causative agent of hand-foot-and-mouth disease (HFMD).
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.4 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
67.4 kDa | |
Human Kremen-1, Fc Tag (KR1-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 21 - Thr 392 (Accession # Q96MU8-1). | |
-20°C | |
KREMEN1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
100 μg | |
Arg 21 - Thr 392 | |
KREMEN, KRM1,Dickkopf receptor/Kringle domain-containing transmembrane protein 1/Kringle-containing protein marking the eye and the nose | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |