Learn More
ACROBiosystems Human Siglec-7 / CD328 Protein, His Tag
MALS verified
Marke:  ACROBiosystems SG7-H5225-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17176701
  • 419.88 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Siglec-7 is a member of the human CD33‐related Siglec receptor. The extracellular region of Siglec‐7 is characterized by an N‐terminal V‐set Ig domain that can bind sialic acid and two C2‐set Ig domains. The cytoplasmic tail of Siglec‐7 has one immune‐receptor tyrosine‐based inhibitory motif (ITIM) and one ITIM‐like motif. Siglec-7 is considered as a sialic acid-dependent immunoreceptor with inhibitory potential and expressed predominantly on human NK cells, monocytes and a small subset of CD8+ T cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.8 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_055200.1
100 μg
Gln 19 - Leu 353
CDw328,D-siglec,A79 membrane protein,p75,Adhesion inhibitory receptor molecule 1, AIRM-1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
38.8 kDa
Human Sigelc-7, His Tag (SG7-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Leu 353 (Accession # Q9Y286-1).
-20°C
Siglec-7
Human
25 mM MES, 150 mM NaCl, pH5.5
HEK293
Bio-Markers and CD Antigens