Learn More
ACROBiosystems Human PSGL-1 / CD162 Protein, His Tag
Marke: ACROBiosystems PS1-H52H9-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
P-selectin glycoprotein ligand 1 (PSGL-1) is also known as Selectin P ligand (SELPLG), CD antigen CD162. PSGL-1 is disulfide-linked homodimer which is the high affinitycounter-receptor for P-selectin on expressed on activated endothelial cells and platelets. PSGL-1 / SELPLG interacts with P-, E- and L-selectins, through their lectin/EGF domains, is required for promoting recruitment and rolling of leukocytes. These interactions require sialyl Lewis X glycan modification but there is a differing dependence for tyrosine sulfations. Dimerization appears not to be required for P-selectin/SELP binding. PSGL-1 can interact with SNX20, MSN and SYK. PSGL-1 / SELPLG mediate the activation of SYK by SELPLG.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.7 kDa. The protein migrates as 55-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002997.2 | |
100 μg | |
Gln 42 - Cys 320 | |
PSGL1,CD162,SELPLG,Selectin P ligand | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
30.7 kDa | |
Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession # Q14242-1). | |
-20°C | |
PSGL-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |