missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,9H,9H-Perfluor-1,9-Nonandiol, Tech. 90 %, Thermo Scientific Chemicals

Catalog Number p-7046859
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
203303-01-9
C9H6F14O2
412.123
MFCD00153275
XQULMKMNFZLURS-UHFFFAOYSA-N
1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol, 1h,1h,9h,9h-perfluorononane-1,9-diol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol
2776243
2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoronan-1,9-Diol
C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
203303-01-9
C9H6F14O2
412.123
MFCD00153275
XQULMKMNFZLURS-UHFFFAOYSA-N
1h,1h,9h,9h-perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro-1,9-nonanediol, 1h,1h,9h,9h-perfluorononane-1,9-diol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonane-1,9-diol
2776243
2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoronan-1,9-Diol
C(C(C(C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1H,1H,9H,9H-Perfluoro-1,9-nonanediol
Schmelzpunkt 108°C to 112°C
Siedepunkt 140°C (5 mmHg)
Strukturformel HOCH2(CF2)7CH2OH
Menge 5 g
Formelmasse 412.12
Reinheit (%) 90%
Güte Eigenschaften

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'