missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-1,2-Cyclohexanedimethanol, TRC

Artikelnummer. 30352564
Change view
Click to view available options
Menge:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30352564 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352564 Supplier Toronto Research Chemicals Supplier No. TRCC98794525MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material (1R,2R)-1,2-Cyclohexanedimethanol
CAS 65376-05-8
Summenformel C8H16O2
Menge 25 mg
Synonym (1R-trans)-1,2-Cyclohexanedimethanol, (1R,2R)-1,2-Bis(hydroxymethyl)cyclohexane
InChI-Formel InChI=1S/C8H16O2/c9-5-7-3-1-2-4-8(7)6-10/h7-10H,1-6H2/t7-,8-/m0/s1
SMILES OC[C@@H]1CCCC[C@H]1CO
IUPAC-Name [(1R,2R)-2-(hydroxymethyl)cyclohexyl]methanol
Molekulargewicht (g/mol) 144.21
Formelmasse 144.115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.