missing translation for 'onlineSavingsMsg'
Learn More

(1 R,3 S)-(-)-3 -(Boc-amino)cyclopentancarbonsäure, 95 %, Thermo Scientific Chemicals

Catalog Number 15419515
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
161660-94-2
C11H18NO4
228.27
MFCD01320858,MFCD01320857
RNJQBGXOSAQQDG-SFYZADRCSA-M
1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
1512526
CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
161660-94-2
C11H18NO4
228.27
MFCD01320858,MFCD01320857
RNJQBGXOSAQQDG-SFYZADRCSA-M
1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
1512526
CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (1R,3S)-(-)-3-(Boc-amino)cyclopentanecarboxylic acid
IUPAC-Name (1R,3S)-3-{[(tert-butoxy)carbonyl]Amino}cyclopentan-1-Carboxylat
Formelmasse 229.28
Reinheit (%) 95%
Menge 250 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'