missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,1,3-Benzothiadiazol-4-yl-isocyanat, 97 %, Maybridge
Click to view available options
Menge:
250 mg
5 g
Spezifikation
Chemischer Name oder Material | 2, 1, 3-Benzothiadiazol-4-yl isocyanate |
CAS Min. % | 96.5 |
CAS Max. % | 100.0 |
Prozentgehaltsbereich | 97% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 250 mg |
Synonym | 2,1,3-benzothiadiazol-4-yl isocyanate, 2,1,3-benzothiadiazol-4-yl isoxyanate, 4-isocyanatobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazol-4-ylisocyanate, 2,1,3-benzothiadiazole,4-isocyanato, benzo c 1,2,5-thiadiazol-4-isocyanate, 2,1,3-benzothiadiazole, 4-isocyanato, 2,1,3-benzothiadiazole, 4-isocyanato-9ci |
Formelmasse | 177.19 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'