missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-benzoesäure, TRC
Specifications
Specifications
| Chemischer Name oder Material | Sodium Cinnamate |
| CAS | 538-42-1 |
| Summenformel | C9 H7 O2 . Na |
| Menge | 500 mg |
| Synonym | 2-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-benzoic Acid, 2-Propenoic acid','3-phenyl-','sodium salt (1:1) (ACI), 2-Propenoic acid','3-phenyl-','sodium salt (9CI), Cinnamic acid','sodium salt (8CI), 3-Phenylpropenoic acid sodium salt, Fortificar, Sodium cinnamate, Sodium β-phenylacrylate, Cinnamic acid sodium salt |
| InChI-Formel | InChI=1S/C9H8O2.Na/c10-9(11)7-6-8-4-2-1-3-5-8;/h1-7H,(H,10,11);/q;+1/p-1 |
| SMILES | [Na+].[O-]C(=O)C=Cc1ccccc1 |
| IUPAC-Name | Natrium;3-Phenylprop-2-enoat |
| Molekulargewicht (g/mol) | 170.141 |
| Formelmasse | 170.034 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?