Learn More
2-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-benzoesäure, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Sodium Cinnamate |
| CAS | 538-42-1 |
| Summenformel | C9 H7 O2 . Na |
| Menge | 1 g |
| Synonym | 2-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-benzoic Acid, 2-Propenoic acid','3-phenyl-','sodium salt (1:1) (ACI), 2-Propenoic acid','3-phenyl-','sodium salt (9CI), Cinnamic acid','sodium salt (8CI), 3-Phenylpropenoic acid sodium salt, Fortificar, Sodium cinnamate, Sodium β-phenylacrylate, Cinnamic acid sodium salt |
| InChI-Formel | InChI=1S/C9H8O2.Na/c10-9(11)7-6-8-4-2-1-3-5-8;/h1-7H,(H,10,11);/q;+1/p-1 |
| SMILES | [Na+].[O-]C(=O)C=Cc1ccccc1 |
| IUPAC-Name | Natrium;3-Phenylprop-2-enoat |
| Molekulargewicht (g/mol) | 170.141 |
| Formelmasse | 170.034 |