missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Difluorphenoxy)-5-Fluor-N-Methylbenzylamin, 96 %, Thermo Scientific™

Catalog Number p-7033588
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
902836-70-8
C14H12F3NO
267.251
MFCD08064232
MFAJRYWQXVCHHW-UHFFFAOYSA-N
2-2,4-difluorophenoxy-5-fluoro-n-methylbenzylamine, 1-2-2,4-difluorophenoxy-5-fluorophenyl-n-methylmethylamine, 2-2,4-difluoro-phenoxy-5-fluoro-benzyl-methyl-amine, 2-2,4-difluorophenoxy-5-fluorophenyl methyl methylamine, 1-2-2,4-difluorophenoxy-5-fluorophenyl-n-methylmethanamine, 2-2,4-difluorophenoxy-5-fluorophenyl methyl methyl amine
24208805
1-[2-(2,4-difluorphenoxy)-5-fluorphenyl]-N-Methylmethanamin
CNCC1=C(C=CC(=C1)F)OC2=C(C=C(C=C2)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
902836-70-8
C14H12F3NO
267.251
MFCD08064232
MFAJRYWQXVCHHW-UHFFFAOYSA-N
2-2,4-difluorophenoxy-5-fluoro-n-methylbenzylamine, 1-2-2,4-difluorophenoxy-5-fluorophenyl-n-methylmethylamine, 2-2,4-difluoro-phenoxy-5-fluoro-benzyl-methyl-amine, 2-2,4-difluorophenoxy-5-fluorophenyl methyl methylamine, 1-2-2,4-difluorophenoxy-5-fluorophenyl-n-methylmethanamine, 2-2,4-difluorophenoxy-5-fluorophenyl methyl methyl amine
24208805
1-[2-(2,4-difluorphenoxy)-5-fluorphenyl]-N-Methylmethanamin
CNCC1=C(C=CC(=C1)F)OC2=C(C=C(C=C2)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(2,4-Difluorophenoxy)-5-fluoro-N-methylbenzylamine
Menge 250 mg
Empfindlichkeit Air sensitive; May form a precipitate
Formelmasse 267.25
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'