Learn More
2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole, TRC
Product Code. 30362413
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362413 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30362413 Supplier Toronto Research Chemicals Supplier No. TRCC3754351G
 Restricted Item
Verfügbar ab: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole
CAS 7189-82-4
Molecular Formula C42 H28 Cl2 N4
Quantity 1 g
Synonym 2-(2-chlorophenyl)-1-[2-(2-chlorophenyl)-4,5-diphenyl-2H-imidazol-2-yl]-4,5-diphenyl-1H-Imidazole, 2-(o-chlorophenyl)-1-[2-(o-chlorophenyl)-4,5-diphenyl-2H-imidazol-2-yl]-4,5-diphenyl-Imidazole (7CI,8CI), 2-(2-Chlorophenyl)-1-[2-(2-chlorophenyl)-4,5-diphenyl-2H-imidazol-2-yl]-4,5-diphenyl-1H-imidazole, 2,2-Bis-(2-chlorophenyl)-4,5,4',5'-tetraphenyl-2'H-[1,2]biimidazolyl, 2,2’-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole, 2,2’-Bis(2-chlorophenyl)-4,5,4',5'-tetraphenyl-1,2'-biimidazole, 2,2’-Bis(o-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole, 2,2’-Bis(o-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-diimidazole, 2,2’-Bis(o-chlorophenyl)-4,5,4',5'-tetraphenyl-1,2'-biimidazole, 2-(2-Chlorophenyl)-1-[2-(2-chlorophenyl)-4,5-diphenyl-1,3-diazol-2-yl]-4,5-diphenylimidazole, BIMD, Speedcure BCIM, o-Cl-HABI
InChI Formula InChI=1S/C42H28Cl2N4/c43-35-27-15-13-25-33(35)41-45-39(31-21-9-3-10-22-31)40(32-23-11-4-12-24-32)48(41)42(34-26-14-16-28-36(34)44)46-37(29-17-5-1-6-18-29)38(47-42)30-19-7-2-8-20-30/h1-28H
SMILES Clc1ccccc1c2nc(c3ccccc3)c(c4ccccc4)n2C5(N=C(C(=N5)c6ccccc6)c7ccccc7)c8ccccc8Cl
IUPAC Name 2-(2-chlorophenyl)-1-[2-(2-chlorophenyl)-4,5-diphenyl-imidazol-2-yl]-4,5-diphenyl-imidazole
Molecular Weight (g/mol) 659.6
Formula Weight 658.1691