missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorbenzolboronsäure-Pinacolester, 96 %, Thermo Scientific Chemicals

Catalog Number p-7040616
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
325142-81-2
C12H12BF5O2
294.028
MFCD12405352
FTJVUPKSBLYGSY-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-perfluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-pentafluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl-1,3,2-dioxaborolane, perfluorophenylboronic acid,pinacol ester, perfluorophenylbornic acid, pinacol ester, perfluorophenylboronic acid, pinacol ester, 2,3,4,5,6-pentafluorobenzeneboronic acid pinacol ester, 2,3,4,5,6-pentafluorophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-2,3,4,5,6-pentafluorobenzene
23065713
4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorphenyl)-1,3,2-dioxaborolan
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C(=C2F)F)F)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
325142-81-2
C12H12BF5O2
294.028
MFCD12405352
FTJVUPKSBLYGSY-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-perfluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-pentafluorophenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl-1,3,2-dioxaborolane, perfluorophenylboronic acid,pinacol ester, perfluorophenylbornic acid, pinacol ester, perfluorophenylboronic acid, pinacol ester, 2,3,4,5,6-pentafluorobenzeneboronic acid pinacol ester, 2,3,4,5,6-pentafluorophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2,3,4,5,6-pentafluorophenyl, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-2,3,4,5,6-pentafluorobenzene
23065713
4,4,5,5-tetramethyl-2-(2,3,4,5,6-pentafluorphenyl)-1,3,2-dioxaborolan
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C(=C2F)F)F)F)F

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,3,4,5,6-Pentafluorobenzeneboronic acid pinacol ester
Schmelzpunkt 35°C to 36°C
Siedepunkt 66°C (0.4 mmHg)
Menge 250 mg
Formelmasse 294.03
Reinheit (%) 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'