missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrahydro-1H-1,4-benzodiazepin, 95 %, Thermo Scientific Chemicals

Catalog Number p-7030283
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5946-39-4
C9H12N2
148.209
MFCD03789577
MLXBHOCKBUILHN-UHFFFAOYSA-N
2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepine
2771762
2,3,4,5-Tetrahydro-1H-1,4-Benzodiazepin
C1CNC2=CC=CC=C2CN1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5946-39-4
C9H12N2
148.209
MFCD03789577
MLXBHOCKBUILHN-UHFFFAOYSA-N
2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepine
2771762
2,3,4,5-Tetrahydro-1H-1,4-Benzodiazepin
C1CNC2=CC=CC=C2CN1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine
Schmelzpunkt 96°C to 100°C
Menge 250 mg
UN-Nummer UN3259
Empfindlichkeit Air Sensitive
Formelmasse 148.21
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'