missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzoesäure, 98+ %, Thermo Scientific Chemicals

Catalog Number p-3779799
Click to view available options
Menge:
5 g
25 g
100 g
Verpackung:
Glasflasche
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
573-11-5
C10H12O5
212.2
MFCD00002433
HZNQSWJZTWOTKM-UHFFFAOYSA-N
benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid
11308
2,3,4-Trimethoxybenzoesäure
COC1=C(C(=C(C=C1)C(=O)O)OC)OC
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
573-11-5
C10H12O5
212.2
MFCD00002433
HZNQSWJZTWOTKM-UHFFFAOYSA-N
benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid
11308
2,3,4-Trimethoxybenzoesäure
COC1=C(C(=C(C=C1)C(=O)O)OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2, 3, 4-Trimethoxybenzoic acid
CAS Min. % 98.0
CAS Max. % 100.0
Schmelzpunkt 99°C to 102°C
Farbe Weiß
Infrarotspektrum Authentic
Prozentgehaltsbereich 98+%
Verpackung Glasflasche
Strukturformel (CH3O)3C6H2CO2H
Menge 5 g
Beilstein 10, 465
Löslichkeitsinformationen Solubility in water: insoluble
Formelmasse 212.2
Reinheit (%) 98+%
Physikalische Form Kristallpulver
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'