Learn More
2,3,5-Tri-O-Benzyl-D-Arabinofuranose, Thermo Scientific Chemicals
Catalog Number p-7043274
Click to view available options
Menge:
1 g
5 g
37776-25-3
C26H28O5
420.505
MFCD00080813
NAQUAXSCBJPECG-DYXQDRAXSA-N
2,3,5-tri-o-benzyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-d-arabinofurannose, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, d-ribose,2,3,5-tris-o-phenylmethyl, 2-o,3-o,5-o-tribenzyl-d-arabinofuranose, 2,3,5-tris-o-phenylmethyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-alpha/beta-d-arabinofuranose, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
11058881
(3S,4R,5R)-3,4-bis(Phenylmethoxy)-5-(Phenylmethoxymethyl)oxolan-2-ol
C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4
37776-25-3
C26H28O5
420.505
MFCD00080813
NAQUAXSCBJPECG-DYXQDRAXSA-N
2,3,5-tri-o-benzyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-d-arabinofurannose, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, d-ribose,2,3,5-tris-o-phenylmethyl, 2-o,3-o,5-o-tribenzyl-d-arabinofuranose, 2,3,5-tris-o-phenylmethyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-alpha/beta-d-arabinofuranose, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
11058881
(3S,4R,5R)-3,4-bis(Phenylmethoxy)-5-(Phenylmethoxymethyl)oxolan-2-ol
C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,3,5-Tri-O-benzyl-D-arabinofuranose
Farbe Weiß
Menge 1 g
Formelmasse 420.5
Physikalische Form Pulver