missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trifluorbenzolsulfonylchlorid, 97 %, Thermo Scientific™

Catalog Number 15465047
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
914636-99-0
C6H2ClF3O2S
230.59
MFCD08532462
UOBDGEAGDFWWOQ-UHFFFAOYSA-N
2,3,5-trifluorobenzenesulphonyl chloride, chloro 2,3,5-trifluorophenyl sulfone, 2,3,5-trifluorophenylsulfonyl chloride, 2,3,5-trifluorobenzene-1-sulfonyl chloride, 2,3,5-trifluorobenzenesulfonylchloride,jrd
44717591
2,3,5-Trifluorbenzolsulfonyl;Chlorid
FC1=CC(F)=C(F)C(=C1)S(Cl)(=O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
914636-99-0
C6H2ClF3O2S
230.59
MFCD08532462
UOBDGEAGDFWWOQ-UHFFFAOYSA-N
2,3,5-trifluorobenzenesulphonyl chloride, chloro 2,3,5-trifluorophenyl sulfone, 2,3,5-trifluorophenylsulfonyl chloride, 2,3,5-trifluorobenzene-1-sulfonyl chloride, 2,3,5-trifluorobenzenesulfonylchloride,jrd
44717591
2,3,5-Trifluorbenzolsulfonyl;Chlorid
FC1=CC(F)=C(F)C(=C1)S(Cl)(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,3,5-Trifluorobenzenesulfonyl chloride
Schmelzpunkt 40°C to 49°C
Menge 1 g
UN-Nummer UN3261
Empfindlichkeit Moisture sensitive
Formelmasse 230.59
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'