missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibrom-6-chlorpyridin, 95 %, Thermo Scientific™

Catalog Number p-7029647
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
885952-16-9
C5H2Br2ClN
271.34
MFCD06798229
YAEPXEMYHIOVFX-UHFFFAOYSA-N
pyridine,2,3-dibromo-6-chloro
42553002
2,3-Dibrom-6-Chlorpyridin
ClC1=NC(Br)=C(Br)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
885952-16-9
C5H2Br2ClN
271.34
MFCD06798229
YAEPXEMYHIOVFX-UHFFFAOYSA-N
pyridine,2,3-dibromo-6-chloro
42553002
2,3-Dibrom-6-Chlorpyridin
ClC1=NC(Br)=C(Br)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,3-Dibromo-6-chloropyridine
Schmelzpunkt 69°C to 71°C
Menge 250 mg
Formelmasse 271.34
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'