missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluor-4-Methoxybenzoesäure, 97+ %, Thermo Scientific™

Catalog Number p-7028713
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
329014-60-0
C8H6F2O3
188.13
MFCD03428569
OBSJPUVORWCLNA-UHFFFAOYSA-N
3611682
2,3-difluor-4-methoxybenzoesäure
COC1=C(C(=C(C=C1)C(=O)O)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
329014-60-0
C8H6F2O3
188.13
MFCD03428569
OBSJPUVORWCLNA-UHFFFAOYSA-N
3611682
2,3-difluor-4-methoxybenzoesäure
COC1=C(C(=C(C=C1)C(=O)O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,3-Difluoro-4-methoxybenzoic acid
Schmelzpunkt 222°C to 224°C
Menge 250 mg
Synonym 2,3-difluoro-4-methoxy-benzoic acid, benzoic acid,2,3-difluoro-4-methoxy, benzoic acid, 2,3-difluoro-4-methoxy, pubchem4939, acmc-20aoh0, ksc569k3p
Formelmasse 188.13
Reinheit (%) ≥97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'