missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrothieno[3,4 -b][1,4]dioxin-5-carbaldehyd, ≥97 %, Thermo Scientific™
Click to view available options
Menge:
250 mg
1 g
5 g
Specifications
Chemischer Name oder Material | 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde |
CAS Min. % | 96.5 |
CAS Max. % | 100.0 |
Prozentgehaltsbereich | 97% |
Verpackung | Bernsteinfarbene Glasflasche |
Menge | 250 mg |
Formelmasse | 170.19 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'