missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)Benzimidazol-6-Carbonsäure, 97%, Thermo Scientific Chemicals

Catalog Number p-7036321
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
312496-16-5
C13H9N3O2
239.234
MFCD01414693
DHRPCXYIVJXXJR-UHFFFAOYSA-N
1h-benzimidazole-6-carboxylicacid, 2-3-pyridinyl, 2-pyridin-3-yl-3h-1,3-benzodiazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzoimidazole-5-carboxylic acid, 2-3-pyridyl benzimidazole-6-carboxylic acid, 6-carboxy-2-pyridin-3-yl-1h-benzimidazole, 2-pyridin-3-yl-1h-benzoimidazole-5-carboxylic acid, 2-3-pyridinyl-1h-benzoimidazole-5-carboxylic acid, 2-pyridin-3-yl-1h-benzimidazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzimidazole-5-carboxylic acid
2771805
2-Pyridin-3-yl-3H-benzimidazol-5-carbonsäure
C1=CC(=CN=C1)C2=NC3=C(N2)C=C(C=C3)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
312496-16-5
C13H9N3O2
239.234
MFCD01414693
DHRPCXYIVJXXJR-UHFFFAOYSA-N
1h-benzimidazole-6-carboxylicacid, 2-3-pyridinyl, 2-pyridin-3-yl-3h-1,3-benzodiazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzoimidazole-5-carboxylic acid, 2-3-pyridyl benzimidazole-6-carboxylic acid, 6-carboxy-2-pyridin-3-yl-1h-benzimidazole, 2-pyridin-3-yl-1h-benzoimidazole-5-carboxylic acid, 2-3-pyridinyl-1h-benzoimidazole-5-carboxylic acid, 2-pyridin-3-yl-1h-benzimidazole-5-carboxylic acid, 2-pyridin-3-yl-3h-benzimidazole-5-carboxylic acid
2771805
2-Pyridin-3-yl-3H-benzimidazol-5-carbonsäure
C1=CC(=CN=C1)C2=NC3=C(N2)C=C(C=C3)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(3-Pyridyl)benzimidazole-6-carboxylic acid
Menge 250 mg
Formelmasse 239.23
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'