missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentanol, TRC

Product Code. 30456529
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30456529 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456529 Supplier Toronto Research Chemicals Supplier No. TRCT79640510MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 2,4,4-Trimethyl-1-pentanol
CAS 16325-63-6
Summenformel C8H18O
Menge 10 mg
Synonym NSC 24283
InChI-Formel InChI=1S/C8H18O/c1-7(6-9)5-8(2,3)4/h7,9H,5-6H2,1-4H3
SMILES CC(CO)CC(C)(C)C
IUPAC-Name 2,4,4-Trimethylpentan-1-ol
Molekulargewicht (g/mol) 130.23
Formelmasse 130.1358
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.