Learn More
2,4,6-Tribromophenyl caproate, MedChemExpress
Product Code. 16490926
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490926 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16490926 Supplier Medchem Express Supplier No. HY101506
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
MedChemExpress 2,4,6-Tribromophenyl caproate (2,4,6-tribromophenyl caproic acid ester) is an anti-fungal agent.
2,4,6-Tribromophenyl caproate (2,4,6-tribromophenyl caproic acid ester) is an anti-fungal agent.
Specifications
Chemical Name or Material 2,4,6-Tribromophenyl caproate
Physical Form Liquid
CAS 16732-09-5
Color Colorless
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C12H13Br3O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (116.57 mM)
SMILES CCCCCC(OC1=C(Br)C=C(Br)C=C1Br)=O
Molecular Weight (g/mol) 428.94
Formula Weight 428.94
Percent Purity 98.29%
Grade Research