missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorphenylacetonitril, 98 %, Thermo Scientific™

Catalog Number p-7024909
missing translation for 'orderingAttributeHoverText'
Menge:
0.5 g
2.5 g
Este artículo no se puede devolver. Vea la política de devoluciones
220227-80-5
C8H4F3N
171.122
MFCD00061221
IHCQMVSMIRAMBR-UHFFFAOYSA-N
2,4,6-trifluorophenylacetonitrile, 2-2,4,6-trifluorophenyl acetonitrile, 2,4,6-trifluorobenzyl cyanide, trifluorophenylacetonitrile 246, 2-2,4,6-trifluorophenyl ethanenitrile, acmc-1ccou, ksc548g0r, 2,4,6-trifluorophenyl acetonitrile, benzeneacetonitrile,2,4,6-trifluoro
2777958
2-(2,4,6-trifluorphenyl)acetonitril
C1=C(C=C(C(=C1F)CC#N)F)F
Este artículo no se puede devolver. Vea la política de devoluciones
220227-80-5
C8H4F3N
171.122
MFCD00061221
IHCQMVSMIRAMBR-UHFFFAOYSA-N
2,4,6-trifluorophenylacetonitrile, 2-2,4,6-trifluorophenyl acetonitrile, 2,4,6-trifluorobenzyl cyanide, trifluorophenylacetonitrile 246, 2-2,4,6-trifluorophenyl ethanenitrile, acmc-1ccou, ksc548g0r, 2,4,6-trifluorophenyl acetonitrile, benzeneacetonitrile,2,4,6-trifluoro
2777958
2-(2,4,6-trifluorphenyl)acetonitril
C1=C(C=C(C(=C1F)CC#N)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Anwendungen
2,4,6-Trifluorphenylacetonitril wird in Agrochemikalien, Pharmazeutika und Farbstoffen verwendet

Hinweise
Von Oxidationsmitteln entfernt aufbewahren Kühl und trocken in einem gut versiegelten Behälter lagern.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,4,6-Trifluorophenylacetonitrile
Brechungsindex 1.4635
Menge 0.5 g
UN-Nummer UN3276
Formelmasse 171.12
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'